Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017019.1 | NP_058715.1 | 4 | 236 | ||
GenBank mRNA | AJ245642.1 | - | 4 | 200 | ||
AJ245643.1 | - | 3 | 175 | |||
D00403.1 | - | 4 | 236 |