Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017043.4 | NP_058739.4 | 9 | 1281 | ||
XM_006234063.2 | XP_006234125.1 | 9 | 1280 | |||
GenBank mRNA | BC081816.1 | - | 9 | 1281 | 7129759 | |
S67721.1 | - | 9 | 1274 | |||
U03388.1 | - | 9 | 1274 | |||
U18060.1 | - | 8 | 1222 |