Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FFAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005306.2 | NP_005297.1 | 1 | 473 | ||
XM_017026709.1 | XP_016882198.1 | 3 | 1012 | |||
XM_017026710.1 | XP_016882199.1 | 2 | 889 | |||
XM_017026711.1 | XP_016882200.1 | 2 | 647 | |||
GenBank mRNA | AB378083.1 | - | 1 | 473 | ||
AX810701.1 | - | 2 | 553 | |||
BC095535.1 | - | 1 | 470 | |||
BC096198.3 | - | 1 | 473 | 40004248 | ||
BC096199.3 | - | 1 | 473 | 40004249 | ||
BC096200.1 | - | 1 | 473 | |||
BC096201.1 | - | 1 | 473 | 40004253 |