Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017060.2 | NP_058756.2 | 4 | 229 | ||
XM_006230687.2 | XP_006230749.1 | 3 | 1098 | |||
GenBank mRNA | AB298805.1 | - | 2 | 148 | ||
CB613004.1 | - | 4 | 229 | |||
FQ224126.1 | - | 4 | 289 |