Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099406.1 | NP_001092876.1 | 7 | 1361 | ||
NM_012319.3 | NP_036451.3 | 8 | 2159 | |||
XM_011525900.1 | XP_011524202.1 | 8 | 2592 | |||
XM_011525901.1 | XP_011524203.1 | 9 | 3276 | |||
GenBank mRNA | AK299102.1 | - | 7 | 1192 | ||
AK301539.1 | - | 7 | 1311 | |||
AK304079.1 | - | 8 | 2083 | |||
BC008317.2 | - | 4 | 728 | |||
BC039498.1 | - | 7 | 1352 | 5552785 | ||
DR762943.1 | - | 3 | 479 | |||
U41060.2 | - | 9 | 1988 |