Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017063.1 | NP_058759.1 | 8 | 905 | ||
GenBank mRNA | L38644.1 | - | 8 | 905 |