Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srd5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017070.3 | NP_058766.2 | 854 | |||
GenBank mRNA | BC127454.1 | - | 855 | 7313862 | ||
EF091857.1 | - | 340 | ||||
J05035.1 | - | 707 | ||||
S81448.1 | - | 727 |