Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017077.2 | NP_058773.1 | 2 | 754 | ||
GenBank mRNA | L09648.1 | - | 2 | 755 |