Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017088.2 | NP_058784.2 | 4 | 390 | ||
GenBank mRNA | AF130987.1 | - | 1 | 26 | ||
BC091156.1 | - | 4 | 394 | |||
BC099763.1 | - | 4 | 390 | 7930350 | ||
U07952.1 | - | 4 | 307 | |||
X74402.1 | - | 4 | 296 |