Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpcal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017356.2 | NP_059052.1 | 5 | 933 | ||
XM_006239914.2 | XP_006239976.1 | 6 | 1540 | |||
XM_006239916.2 | XP_006239978.1 | 5 | 1435 | |||
XM_008764605.1 | XP_008762827.1 | 5 | 1050 | |||
XM_008764606.1 | XP_008762828.1 | 7 | 1650 | |||
GenBank mRNA | BC088759.1 | - | 5 | 933 | 7317905 | |
D13126.1 | - | 5 | 888 | |||
FQ214068.1 | - | 3 | 456 |