Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psg19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270654.1 | NP_001257583.1 | 4 | 1932 | ||
NM_019126.2 | NP_061999.2 | 5 | 2292 | |||
GenBank mRNA | BC097328.1 | - | 4 | 1906 | 7462517 | |
U09815.1 | - | 2292 |