Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF508795.1 | - | 4 | 412 | ||
BC062403.1 | - | 4 | 384 | 5621394 | ||
CK480694.1 | - | 4 | 377 | |||
FQ209590.1 | - | 4 | 395 | |||
FQ218571.1 | - | 4 | 397 | |||
FQ220674.1 | - | 4 | 395 | |||
M15327.1 | - | 4 | 368 | |||
X72792.1 | - | 2 | 198 |