Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021655.2 | NP_067687.2 | 6 | 681 | ||
GenBank mRNA | AF145445.1 | - | 6 | 534 | ||
BC087703.1 | - | 6 | 661 | 7313842 | ||
X06832.1 | - | 6 | 669 |