Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012341.2 | NP_036473.2 | 10 | 1126 | ||
GenBank mRNA | AF120334.1 | - | 10 | 1078 | ||
AF325353.1 | - | 10 | 1055 | |||
AK001548.1 | - | 10 | 1073 | |||
AK001552.1 | - | 10 | 1128 | |||
AK097093.1 | - | 8 | 1934 | |||
AK222861.1 | - | 10 | 1073 | |||
AK295231.1 | - | 9 | 969 | |||
AK301721.1 | - | 10 | 1241 | |||
AK302219.1 | - | 9 | 931 | |||
BC033784.1 | - | 10 | 1049 | |||
BC038975.2 | - | 10 | 1087 | 5534986 |