Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021739.2 | NP_068507.2 | 13, 14 | 1111 | ||
XM_006251170.2 | XP_006251232.1 | 13, 14 | 1210 | |||
XM_006251171.2 | XP_006251233.1 | 13, 14 | 1210 | |||
XM_006251176.2 | XP_006251238.1 | 13, 14 | 1191 | |||
GenBank mRNA | M16112.1 | - | 13, 14 | 1091 |