Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021739.2 | NP_068507.2 | 16 | 1232 | ||
XM_006251170.2 | XP_006251232.1 | 16 | 1331 | |||
XM_006251172.2 | XP_006251234.1 | 15 | 1259 | |||
XM_006251177.2 | XP_006251239.1 | 15 | 1239 | |||
GenBank mRNA | M16112.1 | - | 16 | 1212 |