Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022519.2 | NP_071964.2 | 3 | 435 | ||
XM_006240456.1 | XP_006240518.1 | 2 | 292 | |||
GenBank mRNA | BC078824.1 | - | 3 | 435 | 7131535 | |
D00675.1 | - | 2 | 238 | |||
FQ210655.1 | - | 2 | 228 | |||
FQ218411.1 | - | 2 | 367 | |||
FQ219741.1 | - | 2 | 229 | |||
M32247.1 | - | 1 | 175 |