Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012382.2 | NP_036514.1 | 5 | 704 | ||
XM_011514003.2 | XP_011512305.1 | 5 | 1829 | |||
GenBank mRNA | AF023244.1 | - | 5 | 650 | ||
AK303119.1 | - | 4 | 612 | |||
AK307131.1 | - | 3 | 465 | |||
AK312560.1 | - | 5 | 678 | |||
BC015701.1 | - | 5 | 674 | 3907836 | ||
BC036536.1 | - | 5 | 653 | 5262401 |