Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022666.1 | NP_073157.1 | 1307 | |||
GenBank mRNA | M90518.1 | - | 1307 | |||
M92077.1 | - | 522 | ||||
U47331.1 | - | 1307 |
Set Membership: |
G-protein coupled receptor |