Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012383.4 | NP_036515.4 | 8, 9 | 706 | ||
XM_006717053.3 | XP_006717116.1 | 8, 9 | 715 | |||
XM_006717055.3 | XP_006717118.1 | 7, 8 | 658 | |||
XM_011518524.2 | XP_011516826.1 | 9, 10 | 525 | |||
GenBank mRNA | AK222596.1 | - | 8, 9 | 650 | ||
AK291511.1 | - | 8, 9 | 674 | |||
BC007459.1 | - | 8, 9 | 612 | 4052054 | ||
U63717.1 | - | 8, 9 | 644 |