Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005401.4 | NP_005392.2 | 3, 4 | 997 | ||
XM_017001941.1 | XP_016857430.1 | 4, 5 | 1084 | |||
GenBank mRNA | AK291641.1 | - | 3, 4 | 929 | ||
AK298120.1 | - | 2, 3 | 590 | |||
BC017300.2 | - | 3, 4 | 568 | |||
BC101754.1 | - | 2, 3 | 404 | 8069260 | ||
BC104803.1 | - | 2, 3 | 404 | 8143806 | ||
HQ116786.1 | - | 2, 3 | 344 | |||
X82676.1 | - | 3, 4 | 538 |