Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271814.1 | NP_001258743.1 | 3 | 781 | ||
NM_012400.3 | NP_036532.1 | 4 | 888 | |||
GenBank mRNA | AF188625.1 | - | 4 | 855 | ||
AK290406.1 | - | 4 | 849 | |||
AK310156.1 | - | 3 | 751 | |||
BC025706.1 | - | 4 | 837 | 5223912 |