Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agxt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276706.1 | NP_001263635.1 | 4, 5 | 706 | ||
NM_030656.2 | NP_085914.2 | 4, 5 | 706 | |||
GenBank mRNA | BC088133.1 | - | 4, 5 | 538 | ||
M35270.1 | - | 4, 5 | 706 | |||
X06357.1 | - | 4, 5 | 644 |