Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030838.1 | NP_110465.1 | 15 | 2328 | ||
XM_006237642.2 | XP_006237704.1 | 17 | 2543 | |||
XM_006237643.2 | XP_006237705.1 | 17 | 2563 | |||
XM_006237646.2 | XP_006237708.1 | 15 | 2344 | |||
XM_006237648.2 | XP_006237710.1 | 12 | 2756 | |||
XM_008763432.1 | XP_008761654.1 | 16 | 3912 | |||
GenBank mRNA | AF083469.1 | - | 15 | 2328 | ||
FQ211722.1 | - | 17 | 2558 |