Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270783.1 | NP_001257712.1 | 3, 4 | 314 | ||
NM_001270784.1 | NP_001257713.1 | 3, 4 | 314 | |||
NM_030988.2 | NP_112250.2 | 3, 4 | 314 | |||
GenBank mRNA | AB035201.1 | - | 3, 4 | 314 | ||
AB679745.1 | - | 3, 4 | 288 | |||
AB682738.1 | - | 3, 4 | 288 | |||
AB682739.1 | - | 3, 4 | 288 | |||
AF221622.1 | - | 3, 4 | 288 | |||
M35965.1 | - | 3, 4 | 274 |
Gene Symbol: LOC102550922 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765539.1 | XP_008763761.1 | 1595 |