Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242854.1 | NP_001229783.1 | 5, 6 | 935 | ||
NM_001242855.1 | NP_001229784.1 | 4, 5 | 695 | |||
NM_001242856.1 | NP_001229785.1 | 3, 4 | 576 | |||
NM_012402.3 | NP_036534.1 | 5, 6 | 796 | |||
XM_005252840.4 | XP_005252897.1 | 5, 6 | 700 | |||
GenBank mRNA | AK026390.1 | - | 5, 6 | 628 | ||
AK300855.1 | - | 5, 6 | 796 | |||
AK301856.1 | - | 4, 5 | 650 | |||
AK301972.1 | - | 5, 6 | 737 | |||
AK304512.1 | - | 3, 4 | 443 | |||
AK311004.1 | - | 2 | 364 | |||
BC000392.2 | - | 5, 6 | 618 | 2821666 | ||
U52522.1 | - | 5, 6 | 612 | |||
X97567.1 | - | 3, 4 | 431 |