Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030989.3 | NP_112251.2 | 7 | 946 | ||
XM_006246594.2 | XP_006246656.1 | 8 | 1375 | |||
XM_006246595.2 | XP_006246657.1 | 7 | 1732 | |||
XM_008767773.1 | XP_008765995.1 | 7 | 953 | |||
GenBank mRNA | BC081788.1 | - | 7 | 875 | 7193583 | |
BC098663.1 | - | 7 | 904 | 7384467 | ||
U90328.1 | - | 6 | 815 | |||
X13058.1 | - | 6 | 838 |