Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030989.3 | NP_112251.2 | 8 | 1084 | ||
XM_006246594.2 | XP_006246656.1 | 9 | 1513 | |||
XM_006246595.2 | XP_006246657.1 | 8 | 1870 | |||
XM_008767773.1 | XP_008765995.1 | 8 | 1091 | |||
GenBank mRNA | BC081788.1 | - | 8 | 1013 | 7193583 | |
BC098663.1 | - | 8 | 1042 | 7384467 | ||
FQ232659.1 | - | 8 | 1072 | |||
U90328.1 | - | 7 | 953 | |||
X13058.1 | - | 7 | 976 |