Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030992.1 | NP_112254.1 | 2, 3 | 348 | ||
XM_006232205.2 | XP_006232267.1 | 2, 3 | 1123 | |||
XM_006232206.2 | XP_006232268.1 | 3, 4 | 776 | |||
XM_008760891.1 | XP_008759113.1 | 3, 4 | 776 | |||
GenBank mRNA | AB000778.1 | - | 2, 3 | 363 | ||
AB000779.1 | - | 2, 3 | 348 | |||
AF017251.1 | - | 1, 2 | 192 | |||
U69550.1 | - | 2, 3 | 348 | |||
U88986.1 | - | 2, 3 | 260 |