Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190162.1 | NP_001177091.1 | 4 | 626 | ||
NM_001190163.1 | NP_001177092.1 | 4 | 614 | |||
NM_031511.2 | NP_113699.2 | 4 | 1601 | |||
XM_006230664.2 | XP_006230726.1 | 4 | 1403 | |||
XM_006230665.2 | XP_006230727.1 | 4 | 606 | |||
XM_008760074.1 | XP_008758296.1 | 4 | 1424 | |||
XM_008760075.1 | XP_008758297.1 | 5 | 939 | |||
GenBank mRNA | JQ408382.1 | - | 1539 | |||
M30273.1 | - | 2 | 355 | |||
M38688.1 | - | 3 | 460 | |||
X00911.1 | - | 4 | 1601 | |||
X13101.1 | - | 4 | 638 | |||
X14833.1 | - | 4 | 914 | |||
X14834.1 | - | 4 | 1669 | |||
X16703.1 | - | 1 | 252 |