Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031512.2 | NP_113700.2 | 4 | 226 | ||
GenBank mRNA | BC091141.1 | - | 4 | 226 | 7365029 | |
M98820.1 | - | 4 | 246 |