Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031527.1 | NP_113715.1 | 2 | 223 | ||
GenBank mRNA | BC070517.1 | - | 2 | 203 | 7104674 | |
D00859.1 | - | 2 | 223 | |||
D90163.1 | - | 2 | 207 | |||
FQ224860.1 | - | 2 | 213 | |||
FQ231613.1 | - | 2 | 232 | |||
FQ232496.1 | - | 2 | 231 | |||
FQ233040.1 | - | 2 | 223 | |||
FQ235125.1 | - | 2 | 223 |