Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rara | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031528.2 | NP_113716.2 | 5 | 931 | ||
XM_008767945.1 | XP_008766167.1 | 5 | 731 | |||
GenBank mRNA | AJ002940.1 | - | 5 | 655 | ||
AJ002941.1 | - | 5 | 671 | |||
BC099830.1 | - | 5 | 931 | 7933868 | ||
U15211.1 | - | 5 | 937 |