Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina3n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031531.1 | NP_113719.1 | 2 | 493 | ||
GenBank mRNA | BC078796.1 | - | 2 | 445 | 7129287 | |
D00753.1 | - | 2 | 478 | |||
FQ209856.1 | - | 497 | ||||
FQ210230.1 | - | 2 | 497 | |||
HM448398.1 | - | 389 | ||||
X16359.1 | - | 2 | 493 |