Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012419.4 | NP_036551.3 | 5 | 953 | ||
GenBank mRNA | AF202257.3 | - | 5 | 964 | ||
BC013117.1 | - | 5 | 948 | 3882749 |