Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031770.3 | NP_113958.1 | 1, 2 | 104 | ||
XM_008766344.1 | XP_008764566.1 | 2, 3 | 677 | |||
GenBank mRNA | AF001953.1 | - | 1, 2 | 104 | ||
AF022086.1 | - | 1, 2 | 76 | |||
AY552803.1 | - | 1, 2 | 104 | |||
BC089221.1 | - | 1, 2 | 118 | 7320619 | ||
DQ120491.1 | - | 1, 2 | 111 | |||
DQ120492.1 | - | 1, 2 | 111 |