Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032055.4 | NP_114444.2 | 5 | 1069 | ||
GenBank mRNA | BC101854.1 | - | 5 | 1336 | 7933924 | |
KC222902.1 | - | 5 | 1010 | |||
KC222903.1 | - | 5 | 1010 | |||
KC222904.1 | - | 5 | 1010 | |||
KC222905.1 | - | 5 | 827 | |||
KC222906.1 | - | 5 | 1010 | |||
X57523.1 | - | 5 | 1057 | |||
Y10230.1 | - | 5 | 1010 | |||
Y10231.1 | - | 5 | 1010 | |||
Y10232.1 | - | 5 | 1010 | |||
Y10233.1 | - | 5 | 1010 | |||
Y10234.1 | - | 5 | 1010 | |||
Y10235.1 | - | 5 | 1010 |