Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tapbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033098.2 | NP_149089.1 | 2 | 217 | ||
GenBank mRNA | AJ400732.1 | - | 2 | 206 | ||
BC062007.1 | - | 2 | 208 | 5599850 | ||
CB794019.1 | - | 2 | 276 | |||
FM054278.1 | - | 2 | 208 | |||
KC222917.1 | - | 2 | 194 | |||
KC222918.1 | - | 2 | 194 | |||
KC222919.1 | - | 2 | 194 | |||
KC222920.1 | - | 2 | 194 | |||
KC222921.1 | - | 2 | 194 |