Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hagh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033349.2 | NP_203500.2 | 2, 3 | 285 | ||
XM_006245899.1 | XP_006245961.1 | 2, 3 | 262 | |||
XM_006245900.1 | XP_006245962.1 | 1, 2 | 176 | |||
GenBank mRNA | BC097301.1 | - | 2, 3 | 237 | ||
CB715597.1 | - | 2, 3 | 314 | |||
FQ229806.1 | - | 2, 3 | 354 |