Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052806.2 | NP_434693.1 | 2 | 118 | ||
GenBank mRNA | AY574260.1 | - | 2 | 118 | ||
U42976.1 | - | 178 | ||||
X15834.1 | - | 230 |