Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053290.1 | NP_445742.1 | 4 | 1334 | ||
GenBank mRNA | M76591.1 | - | 4 | 1334 |