Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053543.2 | NP_445995.2 | 7 | 2708 | ||
XM_006238908.1 | XP_006238970.1 | 7 | 2417 | |||
XM_006238909.1 | XP_006238971.1 | 7 | 2478 | |||
GenBank mRNA | BC087000.1 | - | 7 | 2708 | 7190905 |