Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053588.2 | NP_446040.1 | 166 | |||
GenBank mRNA | AF315468.1 | - | 127 | |||
BC061821.1 | - | 166 | 5599087 |