Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1i2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270624.1 | NP_001257553.1 | 7 | 890 | ||
NM_001270625.1 | NP_001257554.1 | 6 | 830 | |||
NM_053880.2 | NP_446332.2 | 8 | 908 | |||
XM_006234344.2 | XP_006234406.1 | 7 | 667 | |||
XM_006234346.2 | XP_006234408.1 | 6 | 606 | |||
GenBank mRNA | BC078764.1 | - | 7 | 636 | 7113840 | |
U39044.1 | - | 8 | 624 | |||
U39045.1 | - | 7 | 606 | |||
U39046.1 | - | 6 | 546 |