Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053996.2 | NP_446448.2 | 2177 | |||
GenBank mRNA | M88111.1 | - | 2176 |