Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cav2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131914.2 | NP_571989.2 | 3 | 738 | ||
GenBank mRNA | BC062059.1 | - | 3 | 738 | 5598269 | |
FQ209775.1 | - | 3 | 792 |