Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133284.2 | NP_579818.1 | 5 | 532 | ||
GenBank mRNA | X04644.1 | - | 5 | 532 |