Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012458.3 | NP_036590.1 | 1, 2 | 513 | ||
GenBank mRNA | AF144700.1 | - | 1, 2 | 176 | ||
AF152352.1 | - | 1, 2 | 130 | |||
AK024764.1 | - | 1, 2 | 148 | |||
BC008607.1 | - | 1, 2 | 171 | 3453921 | ||
BI908100.1 | - | 1, 2 | 474 | |||
DA890574.1 | - | 1, 2 | 485 | |||
DA947460.1 | - | 1, 2 | 520 |