Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012459.2 | NP_036591.2 | 1 | 149 | ||
NR_028383.1 | 1 | 149 | ||||
GenBank mRNA | AF150087.1 | - | 1 | 86 | ||
AF152350.1 | - | 1 | 74 | |||
AF165967.1 | - | 1 | 59 | |||
AK025397.1 | - | 1 | 107 | |||
AK312169.1 | - | 1 | 100 | |||
BC000711.2 | - | 1 | 96 | 2823930 | ||
BC105986.1 | - | 1 | 117 | 4510312 | ||
BC106067.1 | - | 1 | 117 | 4708781 |