Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TINF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099274.1 | NP_001092744.1 | 2, 3 | 644 | ||
NM_012461.2 | NP_036593.2 | 2, 3 | 644 | |||
XM_011536642.2 | XP_011534944.1 | 2, 3 | 616 | |||
GenBank mRNA | AF195512.1 | - | 2, 3 | 564 | ||
AK023166.1 | - | 2, 3 | 585 | |||
AK225379.1 | - | 2, 3 | 588 | |||
AK315084.1 | - | 2, 3 | 532 | |||
BC005030.1 | - | 2, 3 | 425 | 3690254 | ||
BC019343.1 | - | 2, 3 | 602 | 3505009 | ||
BM549897.1 | - | 2, 3 | 627 | |||
BX161478.1 | - | 2, 3 | 577 |